Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-1-methylpyridinium iodide, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
14338-32-0
C6H7ClIN
255.49
MFCD00011984
ABFPKTQEQNICFT-UHFFFAOYSA-M
2-chloro-1-methylpyridinium iodide,2-chloro-1-methylpyridin-1-ium iodide,pyridinium, 2-chloro-1-methyl-, iodide,unii-rlq88nyz68,rlq88nyz68,2-chloro-1-methylpyridiniumiodide,2-chloro-1-methyl-pyridinium iodide,pubchem16659,acmc-1brmd,ksc494o1l
167069
2-chloro-1-methylpyridin-1-ium;iodide
C[N+]1=CC=CC=C1Cl.[I-]
14338-32-0
C6H7ClIN
255.49
MFCD00011984
ABFPKTQEQNICFT-UHFFFAOYSA-M
2-chloro-1-methylpyridinium iodide,2-chloro-1-methylpyridin-1-ium iodide,pyridinium, 2-chloro-1-methyl-, iodide,unii-rlq88nyz68,rlq88nyz68,2-chloro-1-methylpyridiniumiodide,2-chloro-1-methyl-pyridinium iodide,pubchem16659,acmc-1brmd,ksc494o1l
167069
2-chloro-1-methylpyridin-1-ium;iodide
C[N+]1=CC=CC=C1Cl.[I-]
C-X bond formation reagent
Melting Point 195°C to 200°C
Quantity 5g
Infrared Spectrum Authentic
Packaging Glass bottle
Color Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96.0 to 104% (Argentometry)
Fieser 08,95; 12,116
Merck Index 15, 6387
Solubility Information Solubility in water: soluble
Formula Weight 255.49
Physical Form Crystalline Powder and/or Chunks
Percent Purity 97%
Chemical Name or Material 2-Chloro-1-methylpyridinium iodide, 97%
Show More Show Less

GHS H Statement:
Causes skin irritation.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 238-288-7

TSCA : TSCA