Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-1-tritylimidazole, 97%

Manufacturer:  Alfa Aesar™ H51861MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51861MD


  Check Availability
Add to cart

Specifications

Description

Applications
2-Chloro-1-tritylimidazole is used in pharmaceuticals, drug candidates, ligands for transition metal catalysts and other molecular functional materials.

Solubility
Insoluble in water.

Notes
Stable under recommended storage conditions. Store away from oxidizing agents.
Description & Specifications

Chemical Identifiers

67478-48-2
344.84
2-Chloro-1-tritylimidazole
C22H17ClN2
MFCD02179527

Specifications

200° to 202°C
67478-48-2
344.84
2-Chloro-1-tritylimidazole
250mg
C22H17ClN2
MFCD02179527
97%

Safety and Handling

SDS & Safety Documents

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P280g-P305+P351+P338
Wear protective gloves.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

Warning

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit