Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine, 99+%

Click to view available options
Quantity:
100mg
250mg
123318-82-1
C10H11ClFN5O3
303.678
MFCD00871077
WDDPHFBMKLOVOX-AYQXTPAHSA-N
clofarabine,clolar,evoltra,clofarex,cafda,clofarabina,clofarabinum,clofarabine usan,cl-f-ara-a,c1-f-ara-a
119182
CHEBI:681569
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)Cl
123318-82-1
C10H11ClFN5O3
303.678
MFCD00871077
WDDPHFBMKLOVOX-AYQXTPAHSA-N
clofarabine,clolar,evoltra,clofarex,cafda,clofarabina,clofarabinum,clofarabine usan,cl-f-ara-a,c1-f-ara-a
119182
CHEBI:681569
(2R,3R,4S,5R)-5-(6-amino-2-chloropurin-9-yl)-4-fluoro-2-(hydroxymethyl)oxolan-3-ol
C1=NC2=C(N1C3C(C(C(O3)CO)O)F)N=C(N=C2N)Cl
  • A purine nucleoside anti-metabolite
  • Toxic to nondividing lymphocytes and monocytes
Color White
Quantity 100mg
UN Number UN2811
Merck Index 14,2372
Formula Weight 303.68
Physical Form Powder
Percent Purity ≥99%
Chemical Name or Material 2-Chloro-2'-arabino-fluoro-2'-deoxyadenosine

GHS H Statement
H301
Toxic if swallowed.

GHS P Statement
P264-P270-P301+P310-P321-P405-P501a
Wash thoroughly after handling.
Do not eat, drink or smoke when using this product.
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
Specific treatment (see label).
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: TOXIC SOLIDS, ORGANIC, N.O.S.

RTECSNumber : UD7473000

Recommended Storage : 2° to 8°C