Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3,4-dimethoxybenzaldehyde, 98%, ACROS Organics™

Click to view available options
Quantity:
25g
Packaging:
Glass bottle
5417-17-4
C9H9ClO3
200.618
MFCD00084942
SAWHDJTZESXNMM-UHFFFAOYSA-N
2-chloroveratraldehyde,2-chloro-3,4-dimethoxy-benzaldehyde,benzaldehyde, 2-chloro-3,4-dimethoxy,2-chloro-3,4-dimethoxy benzaldehyde,chloroveratraldehyde,pubchem7759,acmc-1aumn,2-chloro-3,4 dimethoxybenzaldehyde,benzaldehyde,2-chloro-3,4-dimethoxy,2-chloro-3,4-dimethoxybenzaldehyde
79445
2-chloro-3,4-dimethoxybenzaldehyde
COC1=C(C(=C(C=C1)C=O)Cl)OC
5417-17-4
C9H9ClO3
200.618
MFCD00084942
SAWHDJTZESXNMM-UHFFFAOYSA-N
2-chloroveratraldehyde,2-chloro-3,4-dimethoxy-benzaldehyde,benzaldehyde, 2-chloro-3,4-dimethoxy,2-chloro-3,4-dimethoxy benzaldehyde,chloroveratraldehyde,pubchem7759,acmc-1aumn,2-chloro-3,4 dimethoxybenzaldehyde,benzaldehyde,2-chloro-3,4-dimethoxy,2-chloro-3,4-dimethoxybenzaldehyde
79445
2-chloro-3,4-dimethoxybenzaldehyde
COC1=C(C(=C(C=C1)C=O)Cl)OC
Melting Point 69°C to 73°C
Color Beige to Beige-Yellow
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula (CH3O)2C6H2(Cl)CHO
Formula Weight 200.62
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-3, 4-dimethoxybenzaldehyde, 98%
Show More Show Less

GHS H Statement:
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.