Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloro-3,5-dinitropyridine, 98+%, Acros Organics™
Click to view available options
Quantity:
25g
Packaging:
Glass bottle
Specifications
Packaging | Glass bottle |
Melting Point | 63.0°C to 66.0°C |
Color | Yellow |
Infrared Spectrum | Authentic |
Quantity | 25g |
CAS Min % | 98.5 |
CAS Max % | 100.0 |
Assay Percent Range | 98% min. (GC) |
Linear Formula | (O2H)(C5H2N)Cl |
Beilstein | 20,III,2536 |
Show More |
Safety and Handling
GHS H Statement
Fatal if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
GHS P Statement
IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attention.
Wear protective gloves/protective clothing
GHS Signal Word: Danger
EINECSNumber : 219-937-3