Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3,6-difluorobenzaldehyde, 97%

Click to view available options
Quantity:
1g
5g
261762-39-4
C7H3ClF2O
176.547
MFCD01631443
KPYBGKSDBNMHTO-UHFFFAOYSA-N
benzaldehyde, 2-chloro-3,6-difluoro,2,5-difluoro-6-chlorobenzaldehyde,6-chloro-2,5-difluoro-benzaldehyde,pubchem2618,zlchem 1271,acmc-1cfb6,intermediates-zcf02699,ksc495g2n,2-chloro-3,6-difluoro-benzaldehyde,2-chloro-3,6-difluorobenzaldehyde
2773509
2-chloro-3,6-difluorobenzaldehyde
C1=CC(=C(C(=C1F)C=O)Cl)F
261762-39-4
C7H3ClF2O
176.547
MFCD01631443
KPYBGKSDBNMHTO-UHFFFAOYSA-N
benzaldehyde, 2-chloro-3,6-difluoro,2,5-difluoro-6-chlorobenzaldehyde,6-chloro-2,5-difluoro-benzaldehyde,pubchem2618,zlchem 1271,acmc-1cfb6,intermediates-zcf02699,ksc495g2n,2-chloro-3,6-difluoro-benzaldehyde,2-chloro-3,6-difluorobenzaldehyde
2773509
2-chloro-3,6-difluorobenzaldehyde
C1=CC(=C(C(=C1F)C=O)Cl)F
Melting Point 46°C to 48°C
Quantity 1g
Formula Weight 176.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzaldehyde

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning