Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3,6-difluorobenzamide, 97+%

Click to view available options
Quantity:
1g
5g
261762-40-7
C7H4ClF2NO
191.562
MFCD01631444
MDFUJEDMGQUCBB-UHFFFAOYSA-N
acmc-1ch3m,benzamide,2-chloro-3,6-difluoro
2773514
2-chloro-3,6-difluorobenzamide
C1=CC(=C(C(=C1F)C(=O)N)Cl)F
261762-40-7
C7H4ClF2NO
191.562
MFCD01631444
MDFUJEDMGQUCBB-UHFFFAOYSA-N
acmc-1ch3m,benzamide,2-chloro-3,6-difluoro
2773514
2-chloro-3,6-difluorobenzamide
C1=CC(=C(C(=C1F)C(=O)N)Cl)F
Quantity 1g
Formula Weight 191.56
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzamide

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning