Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3,6-difluorobenzoic acid, 97%

Click to view available options
Quantity:
1g
5g
287172-74-1
C7H3ClF2O2
192.546
MFCD01631441
COWJYTLVSFKOPM-UHFFFAOYSA-N
2-chloro-3,6-difluoro-benzoic acid,benzoic acid, 2-chloro-3,6-difluoro,pubchem8530,acmc-1cr2e,benzoic acid,2-chloro-3,6-difluoro,2-chloro-3,6-difluorobenzoic acid
2773530
2-chloro-3,6-difluorobenzoic acid
C1=CC(=C(C(=C1F)C(=O)O)Cl)F
287172-74-1
C7H3ClF2O2
192.546
MFCD01631441
COWJYTLVSFKOPM-UHFFFAOYSA-N
2-chloro-3,6-difluoro-benzoic acid,benzoic acid, 2-chloro-3,6-difluoro,pubchem8530,acmc-1cr2e,benzoic acid,2-chloro-3,6-difluoro,2-chloro-3,6-difluorobenzoic acid
2773530
2-chloro-3,6-difluorobenzoic acid
C1=CC(=C(C(=C1F)C(=O)O)Cl)F
Melting Point 126°C to 128°C
Quantity 5g
Formula Weight 192.55
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzoic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning