Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3,6-difluorobenzyl bromide, 97%

Click to view available options
Quantity:
1g
5g
90292-67-4
C7H4BrClF2
241.46
MFCD01631446
KAADKOQBKOGBAA-UHFFFAOYSA-N
2-chloro-3,6-difluorobenzyl bromide,2-bromomethyl-3-chloro-1,4-difluorobenzene,alpha-bromo-2-chloro-3,6-difluorotoluene,chloro-3,6-difluorobenzylbr,2-chloro-3,6-difluorobenzylbromide,2-chloro 3,6-difluoro-benzyl bromide,2-chloro-3,6-di-fluoro-benzyl bromide,currency-bromo-2-chloro-3,6-difluorotoluene,benzene,2-bromomethyl-3-chloro-1,4-difluoro
2773549
2-(bromomethyl)-3-chloro-1,4-difluorobenzene
C1=CC(=C(C(=C1F)CBr)Cl)F
90292-67-4
C7H4BrClF2
241.46
MFCD01631446
KAADKOQBKOGBAA-UHFFFAOYSA-N
2-chloro-3,6-difluorobenzyl bromide,2-bromomethyl-3-chloro-1,4-difluorobenzene,alpha-bromo-2-chloro-3,6-difluorotoluene,chloro-3,6-difluorobenzylbr,2-chloro-3,6-difluorobenzylbromide,2-chloro 3,6-difluoro-benzyl bromide,2-chloro-3,6-di-fluoro-benzyl bromide,currency-bromo-2-chloro-3,6-difluorotoluene,benzene,2-bromomethyl-3-chloro-1,4-difluoro
2773549
2-(bromomethyl)-3-chloro-1,4-difluorobenzene
C1=CC(=C(C(=C1F)CBr)Cl)F
Quantity 1g
UN Number UN3265
Formula Weight 241.46
Percent Purity 97%
Chemical Name or Material 2-Chloro-3,6-difluorobenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P280-P305+P351+P338-P309-P310
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.