Alfa Aesar™ 2-Chloro-3-cyano-4,6-dimethylpyridine, 98%

Manufacturer: Alfa Aesar™ A1156222

Quantity 100g

 View more versions of this product

Catalog No. AAA1156222

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 14237-71-9
    Molecular Formula C8H7ClN2
    Formula Weight 166.61
    MDL Number MFCD00051676
    UN Number UN3439
    Beilstein 131348
    Synonym 2-Chloro-4,6-dimethylnicotinonitrile; 2-Chloro-4,6-dimethylpyridine-3-carbonitrile
    Chemical Name or Material 2-Chloro-3-cyano-4,6-dimethylpyridine

    Specifications

    Formula Weight 166.61
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Molecular Formula C8H7ClN2
    MDL Number MFCD00051676
    CAS 14237-71-9
    Quantity 100g
    UN Number UN3439
    Synonym 2-Chloro-4,6-dimethylnicotinonitrile; 2-Chloro-4,6-dimethylpyridine-3-carbonitrile
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 97° to 100°C
    Beilstein 131348
    Chemical Name or Material 2-Chloro-3-cyano-4,6-dimethylpyridine
    Assay 98%

    GHS H Statement
    H331-H302-H312-H315-H319-H335
    Toxic if inhaled.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit