Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-cyanopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
25g
Packaging:
Glass bottle
6602-54-6
C6H3ClN2
138.56
MFCD00014628
JAUPUQRPBNDMDT-UHFFFAOYSA-N
2-chloro-3-cyanopyridine,2-chloronicotinonitrile,2-chloro-3-pyridinecarbonitrile,3-cyano-2-chloropyridine,3-pyridinecarbonitrile, 2-chloro,2-chloro-nicotinonitrile,2-chloronicotinitrile,2-chloro-3-cyano-pyridine,2-chloro-3-cyanopyridine 2-chloronicotinonitrile,2chloronicotinonitrile
81079
2-chloropyridine-3-carbonitrile
C1=CC(=C(N=C1)Cl)C#N
6602-54-6
C6H3ClN2
138.56
MFCD00014628
JAUPUQRPBNDMDT-UHFFFAOYSA-N
2-chloro-3-cyanopyridine,2-chloronicotinonitrile,2-chloro-3-pyridinecarbonitrile,3-cyano-2-chloropyridine,3-pyridinecarbonitrile, 2-chloro,2-chloro-nicotinonitrile,2-chloronicotinitrile,2-chloro-3-cyano-pyridine,2-chloro-3-cyanopyridine 2-chloronicotinonitrile,2chloronicotinonitrile
81079
2-chloropyridine-3-carbonitrile
C1=CC(=C(N=C1)Cl)C#N
Infrared Spectrum Authentic
Quantity 25g
Packaging Glass bottle
Color White
Melting Point 107.0°C to 109.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 138.56
Physical Form Crystalline Powder or Flakes
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-cyanopyridine
Show More Show Less

GHS H Statement:
Harmful if inhaled.
Causes skin irritation.
Causes serious eye irritation.
Harmful in contact with skin.
Harmful if swallowed.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

EINECSNumber : 229-550-1