Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
63212-53-3
C9H9BClF4NO
269.431
MFCD00011943
MLQZANYFUYLJRK-UHFFFAOYSA-N
2-chloro-3-ethylbenzoxazolium tetrafluoroborate,2-chloro-3-ethylbenzo d oxazol-3-ium tetrafluoroborate,2-chloro-3-ethyl-1,3-benzoxazol-3-ium tetrafluoroborate,2-chloro-3-ethyl-benzooxazole; tetrafluoroboron,acmc-209ndy,3-ethyl-2-chlorobenzoxazolium tetrafluoroborate,2-chloro-3-ethylbenzoxazo-lium tetrafluoroborate
2724240
2-chloro-3-ethyl-1,3-benzoxazol-3-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC[N+]1=C(OC2=CC=CC=C21)Cl
63212-53-3
C9H9BClF4NO
269.431
MFCD00011943
MLQZANYFUYLJRK-UHFFFAOYSA-N
2-chloro-3-ethylbenzoxazolium tetrafluoroborate,2-chloro-3-ethylbenzo d oxazol-3-ium tetrafluoroborate,2-chloro-3-ethyl-1,3-benzoxazol-3-ium tetrafluoroborate,2-chloro-3-ethyl-benzooxazole; tetrafluoroboron,acmc-209ndy,3-ethyl-2-chlorobenzoxazolium tetrafluoroborate,2-chloro-3-ethylbenzoxazo-lium tetrafluoroborate
2724240
2-chloro-3-ethyl-1,3-benzoxazol-3-ium;tetrafluoroborate
[B-](F)(F)(F)F.CC[N+]1=C(OC2=CC=CC=C21)Cl
Infrared Spectrum Authentic
Packaging Glass bottle
Color Beige
Quantity 1g
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 269.42
Physical Form Crystals or Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Chloro-3-ethylbenzoxazolium tetrafluoroborate, 97%
Show More Show Less

GHS H Statement:
Causes serious eye damage.
Causes severe skin burns and eye damage.

GHS P Statement:
IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger

EINECSNumber : 263-998-9

TSCA : TSCA