Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3-fluoro-6-methylpyridine, 95%

Click to view available options
Quantity:
250mg
1g
374633-32-6
C6H5ClFN
145.561
MFCD03095216
BGDYLOGCMJAUHR-UHFFFAOYSA-N
2-chloro-3-fluoro-6-picoline,abbypharma ap-15-5354,2-chloro-3-fluoro-6-methyl-pyridine,pyridine, 2-chloro-3-fluoro-6-methyl,pubchem1137,2-chloro-3-fluoro-6-methyl pyridine,4-amino-3'-chlorobiphenyl hydrochloride,4-amino-3'-chlorobiphenyl hydrochloride;,2-chloranyl-3-fluoranyl-6-methyl-pyridine,pyridine, 2-chloro-3-fluoro-6-methyl-9ci
40418623
2-chloro-3-fluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)Cl
374633-32-6
C6H5ClFN
145.561
MFCD03095216
BGDYLOGCMJAUHR-UHFFFAOYSA-N
2-chloro-3-fluoro-6-picoline,abbypharma ap-15-5354,2-chloro-3-fluoro-6-methyl-pyridine,pyridine, 2-chloro-3-fluoro-6-methyl,pubchem1137,2-chloro-3-fluoro-6-methyl pyridine,4-amino-3'-chlorobiphenyl hydrochloride,4-amino-3'-chlorobiphenyl hydrochloride;,2-chloranyl-3-fluoranyl-6-methyl-pyridine,pyridine, 2-chloro-3-fluoro-6-methyl-9ci
40418623
2-chloro-3-fluoro-6-methylpyridine
CC1=NC(=C(C=C1)F)Cl
Quantity 1g
Beilstein 22770804
Formula Weight 145.56
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-fluoro-6-methylpyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning