Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-fluoropyridine, 97%, Acros Organics

Click to view available options
Quantity:
5g
25g
17282-04-1
C5H3ClFN
131.54
SVAZIMBLBHOVIR-UHFFFAOYSA-N
3-fluoro-2-chloropyridine,2-chloro-3-fluoro pyridine,pyridine, 2-chloro-3-fluoro,2-chloro-3-fluoro-pyridine,pubchem2532,acmc-1cc0d,ksc174s3h,2-chloro-3-fluoropyridine,2-chloro-3-fluoropyridine 1g
2782791
2-chloro-3-fluoropyridine
C1=CC(=C(N=C1)Cl)F
17282-04-1
C5H3ClFN
131.54
SVAZIMBLBHOVIR-UHFFFAOYSA-N
3-fluoro-2-chloropyridine,2-chloro-3-fluoro pyridine,pyridine, 2-chloro-3-fluoro,2-chloro-3-fluoro-pyridine,pubchem2532,acmc-1cc0d,ksc174s3h,2-chloro-3-fluoropyridine,2-chloro-3-fluoropyridine 1g
2782791
2-chloro-3-fluoropyridine
C1=CC(=C(N=C1)Cl)F
Boiling Point 80°C (80.0mmHg)
Flash Point 63°C
Specific Gravity 1.323
Density 1.3230g/mL
Packaging Glass bottle
Quantity 5g
Assay Percent Range 96% min. (GC)
Formula Weight 131.54
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-fluoropyridine

GHS Signal Word: Warning