Alfa Aesar™ 2-Chloro-3-methylquinoline, 97%

Manufacturer: Alfa Aesar™ H5049206

Quantity 5g

 View more versions of this product

Catalog No. AAH5049206

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 57876-69-4
    Molecular Formula C10H8ClN
    Formula Weight 177.63
    MDL Number MFCD00100265
    Chemical Name or Material 2-Chloro-3-methylquinoline

    Specifications

    Formula Weight 177.63
    Molecular Formula C10H8ClN
    MDL Number MFCD00100265
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Quantity 5g
    CAS 57876-69-4
    Health Hazard 1 Warning
    Health Hazard 3 GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 82° to 84°C
    Chemical Name or Material 2-Chloro-3-methylquinoline
    Assay 97%

    Description

    Applications
    2-Chloro-3-methylquinoline is used as organic chemical synthesis intermediate.

    Solubility
    Insoluble in water.

    Notes
    Stable under recommended storage conditions. Incompatible with oxidizing agents.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P280g-P305+P351+P338
    Wear protective gloves.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit