Alfa Aesar™ 2-Chloro-3-methylthiophene, 97%

Manufacturer: Alfa Aesar™ H2795706

Quantity 5g

 View more versions of this product

Catalog No. AAH2795706

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 14345-97-2
    Molecular Formula C5H5ClS
    Formula Weight 132.61
    MDL Number MFCD00130083
    UN Number UN1993
    Chemical Name or Material 2-Chloro-3-methylthiophene

    Specifications

    Formula Weight 132.61
    Quantity 5g
    Molecular Formula C5H5ClS
    MDL Number MFCD00130083
    CAS 14345-97-2
    UN Number UN1993
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H301-H311-H226-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.
    Health Hazard 3 GHS P Statement
    P210-P301+P310-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 3; Packing Group: III; Proper Shipping Name: FLAMMABLE LIQUIDS, N.O.S.
    Flash Point 46°C (115°F)
    Boiling Point 147° to 149°C
    Chemical Name or Material 2-Chloro-3-methylthiophene
    Assay 97%

    GHS H Statement
    H301-H311-H226-H332
    Toxic if swallowed.
    Toxic in contact with skin.
    Flammable liquid and vapor.
    Harmful if inhaled.

    GHS P Statement
    P210-P301+P310-P303+P361+P353-P361-P405-P501a
    Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
    IF SWALLOWED: Immediately call a POISON CENTER or doctor/physician.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    Remove/Take off immediately all contaminated clothing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit