Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-nitro-4-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
100mg
1g
5g
Packaging:
Glass bottle
23056-39-5
C6H5ClN2O2
172.57
MFCD00012347
JHARVUVBTAAPLA-UHFFFAOYSA-N
2-chloro-3-nitro-4-picoline,pyridine, 2-chloro-4-methyl-3-nitro,2-chloro-3-nitro-4-methylpyridine,2-chloro-4-methyl-3-nitro-pyridine,pubchem6148,acmc-1cui4,2-chloro-3-nitro4-picoline,ksc496c3n,2-chlor-4-methyl-3-nitropyridin
345363
2-chloro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
23056-39-5
C6H5ClN2O2
172.57
MFCD00012347
JHARVUVBTAAPLA-UHFFFAOYSA-N
2-chloro-3-nitro-4-picoline,pyridine, 2-chloro-4-methyl-3-nitro,2-chloro-3-nitro-4-methylpyridine,2-chloro-4-methyl-3-nitro-pyridine,pubchem6148,acmc-1cui4,2-chloro-3-nitro4-picoline,ksc496c3n,2-chlor-4-methyl-3-nitropyridin
345363
2-chloro-4-methyl-3-nitropyridine
CC1=C(C(=NC=C1)Cl)[N+](=O)[O-]
Melting Point 51°C to 53°C
Flash Point 113°C
Infrared Spectrum Authentic
Quantity 1g
Color Yellow to Beige
Packaging Glass bottle
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 97.5% min. (GC)
Formula Weight 172.57
Physical Form Powder
Percent Purity 98%
Chemical Name or Material 2-Chloro-3-nitro-4-picoline
Show More Show Less

GHS H Statement:
Causes skin irritation.
Harmful if swallowed.
Harmful in contact with skin.
May cause respiratory irritation.
Causes serious eye irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning