Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-3-nitro-5-picoline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
23056-40-8
C6H5ClN2O2
172.568
MFCD02070020
LUAJUWOJEFFNFE-UHFFFAOYSA-N
2-chloro-3-nitro-5-picoline,2-chloro-3-nitro-5-methylpyridine,2-chloro-5-methyl-3-nitro-pyridine,2-chloro-5-methyl-3-nitro pyridine,pyridine, 2-chloro-5-methyl-3-nitro,pubchem6171,acmc-1ciox,ksc496c3p,6-chloro-5-nitro-3-picoline
818300
2-chloro-5-methyl-3-nitropyridine
CC1=CC(=C(N=C1)Cl)[N+](=O)[O-]
23056-40-8
C6H5ClN2O2
172.568
MFCD02070020
LUAJUWOJEFFNFE-UHFFFAOYSA-N
2-chloro-3-nitro-5-picoline,2-chloro-3-nitro-5-methylpyridine,2-chloro-5-methyl-3-nitro-pyridine,2-chloro-5-methyl-3-nitro pyridine,pyridine, 2-chloro-5-methyl-3-nitro,pubchem6171,acmc-1ciox,ksc496c3p,6-chloro-5-nitro-3-picoline
818300
2-chloro-5-methyl-3-nitropyridine
CC1=CC(=C(N=C1)Cl)[N+](=O)[O-]
Packaging Glass bottle
Color Beige
Quantity 1g
Melting Point 46°C to 50°C
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 172.57
Physical Form Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-3-nitro-5-picoline
Show More Show Less

GHS H Statement:
Harmful in contact with skin.
Causes skin irritation.
Harmful if swallowed.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.