Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine, 95%

Click to view available options
Quantity:
1g
5g
25g
72587-15-6
C6H2ClF3N2O2
226.539
MFCD06255171
DRPIKFKCAJGTJF-UHFFFAOYSA-N
2-chloro-3-nitro-5-trifluoromethyl pyridine,2-chloro-5-trifluoromethyl-3-nitropyridine,pyridine, 2-chloro-3-nitro-5-trifluoromethyl,5-trifluoromethyl-3-nitro-2-chloropyridine,pubchem17529,acmc-209onn,abbypharma ap-11-5568,2-chloro-3-nitro-5-trifluoromethyl pyridine,2-chloro-3-nitro-5-trifluoromethyl-pyridine,2-chloro3-nitro-5-trifluoromethyl pyridine
4736950
2-chloro-3-nitro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F
72587-15-6
C6H2ClF3N2O2
226.539
MFCD06255171
DRPIKFKCAJGTJF-UHFFFAOYSA-N
2-chloro-3-nitro-5-trifluoromethyl pyridine,2-chloro-5-trifluoromethyl-3-nitropyridine,pyridine, 2-chloro-3-nitro-5-trifluoromethyl,5-trifluoromethyl-3-nitro-2-chloropyridine,pubchem17529,acmc-209onn,abbypharma ap-11-5568,2-chloro-3-nitro-5-trifluoromethyl pyridine,2-chloro-3-nitro-5-trifluoromethyl-pyridine,2-chloro3-nitro-5-trifluoromethyl pyridine
4736950
2-chloro-3-nitro-5-(trifluoromethyl)pyridine
C1=C(C=NC(=C1[N+](=O)[O-])Cl)C(F)(F)F
Color Colorless to Yellow
Quantity 1g
Beilstein 7586778
Formula Weight 226.54
Physical Form Liquid
Percent Purity 95%
Chemical Name or Material 2-Chloro-3-nitro-5-(trifluoromethyl)pyridine

GHS H Statement
H302-H315-H319-H335
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses, if present and easy to do.
Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning