Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3-pyridinemethanol, 97%

Click to view available options
Quantity:
1g
5g
42330-59-6
C6H6ClNO
143.57
MFCD01935961
HMPDWSBKPCOQDW-UHFFFAOYSA-N
2-chloropyridin-3-yl methanol,2-chloro-3-pyridinyl methanol,2-chloro-3-pyridinylmethanol,2-chloro-3-pyridinemethanol,2-chloro-3-hydroxymethyl pyridine,2-chloro-pyridin-3-yl-methanol,3-pyridinemethanol, 2-chloro,2-chloro-3-hydroxymethylpyridine
2763647
(2-chloropyridin-3-yl)methanol
C1=CC(=C(N=C1)Cl)CO
42330-59-6
C6H6ClNO
143.57
MFCD01935961
HMPDWSBKPCOQDW-UHFFFAOYSA-N
2-chloropyridin-3-yl methanol,2-chloro-3-pyridinyl methanol,2-chloro-3-pyridinylmethanol,2-chloro-3-pyridinemethanol,2-chloro-3-hydroxymethyl pyridine,2-chloro-pyridin-3-yl-methanol,3-pyridinemethanol, 2-chloro,2-chloro-3-hydroxymethylpyridine
2763647
(2-chloropyridin-3-yl)methanol
C1=CC(=C(N=C1)Cl)CO
Quantity 5g
Melting Point 61°C to 65°C (decomposition)
Boiling Point 112°C (0.2mmHg)
Beilstein 1525378
Formula Weight 143.57
Physical Form Solid
Percent Purity 97%
Chemical Name or Material 2-Chloro-3-pyridinemethanol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c

H302+H312+H332-H315-H319-H335

TSCA : No

storageNote1 : 2° to 8°C

Recommended Storage : Keep cold