Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-3-(trifluoromethyl)benzyl alcohol, 97+%

Click to view available options
Quantity:
1g
5g
261763-20-6
C8H6ClF3O
210.58
MFCD01631537
MQUXXVLMXCHGDZ-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzyl alcohol,2-chloro-3-trifluoromethyl phenyl methanol,2-chloro-3-trifluoromethyl benzylalcohol,2-chloro-3-trifluoromethyl phenyl methan-1-ol,acmc-1coey,rarechem al bd 0475,rarechem al bd 1301,2-chloro-3-trifluoromethyl-benzylalcohol,2-chloro-3-trifluoromethylbenzyl alcohol
2778108
[2-chloro-3-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C(=C1)C(F)(F)F)Cl)CO
261763-20-6
C8H6ClF3O
210.58
MFCD01631537
MQUXXVLMXCHGDZ-UHFFFAOYSA-N
2-chloro-3-trifluoromethyl benzyl alcohol,2-chloro-3-trifluoromethyl phenyl methanol,2-chloro-3-trifluoromethyl benzylalcohol,2-chloro-3-trifluoromethyl phenyl methan-1-ol,acmc-1coey,rarechem al bd 0475,rarechem al bd 1301,2-chloro-3-trifluoromethyl-benzylalcohol,2-chloro-3-trifluoromethylbenzyl alcohol
2778108
[2-chloro-3-(trifluoromethyl)phenyl]methanol
C1=CC(=C(C(=C1)C(F)(F)F)Cl)CO
Melting Point 74°C to 77°C
Quantity 5g
Formula Weight 210.58
Percent Purity ≥97%
Chemical Name or Material 2-Chloro-3-(trifluoromethyl)benzyl alcohol

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning