Alfa Aesar™ 2-Chloro-4,5-difluorobenzoic acid, 98%

Manufacturer: Alfa Aesar™ B2417414

Quantity 25g

 View more versions of this product

Catalog No. AAB2417414

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 110877-64-0
    Molecular Formula C7H3ClF2O2
    Formula Weight 192.55
    MDL Number MFCD00077479
    Beilstein 4247522
    Chemical Name or Material 2-Chloro-4,5-difluorobenzoic acid

    Specifications

    Formula Weight 192.55
    Molecular Formula C7H3ClF2O2
    MDL Number MFCD00077479
    CAS 110877-64-0
    Beilstein 4247522
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318-H302-H312-H317
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P310-P301+P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 257° to 259°C
    Melting Point 104° to 107°C
    Quantity 25g
    Chemical Name or Material 2-Chloro-4,5-difluorobenzoic acid
    Assay 98%

    GHS H Statement
    H318-H302-H312-H317
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    May cause an allergic skin reaction.

    GHS P Statement
    P261-P280-P305+P351+P338-P310-P301+P312-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit