Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4,6-dimethoxy-1,3,5-triazine, 98%

Click to view available options
Quantity:
5g
25g
100g
3140-73-6
C5H6ClN3O2
175.572
MFCD00075607
GPIQOFWTZXXOOV-UHFFFAOYSA-N
cdmt,1,3,5-triazine, 2-chloro-4,6-dimethoxy,2-chloro-4,6-dimethoxy-s-triazine,2-chloro-4,6-dimethoxy-1,3,5 triazine,pubchem20734,acmc-1cnh7,ksc567a8j,2-chloro-4,6-dimethoxy s-triazine,1,5-triazine, 2-chloro-4,6-dimethoxy
18450
2-chloro-4,6-dimethoxy-1,3,5-triazine
COC1=NC(=NC(=N1)Cl)OC
3140-73-6
C5H6ClN3O2
175.572
MFCD00075607
GPIQOFWTZXXOOV-UHFFFAOYSA-N
cdmt,1,3,5-triazine, 2-chloro-4,6-dimethoxy,2-chloro-4,6-dimethoxy-s-triazine,2-chloro-4,6-dimethoxy-1,3,5 triazine,pubchem20734,acmc-1cnh7,ksc567a8j,2-chloro-4,6-dimethoxy s-triazine,1,5-triazine, 2-chloro-4,6-dimethoxy
18450
2-chloro-4,6-dimethoxy-1,3,5-triazine
COC1=NC(=NC(=N1)Cl)OC
Applications
2-Chloro-4,6-dimethoxy-1,3,5-triazine is used as a peptide coupling agent for the purification of peptides. It is used in the synthesis of 4-(4,6-dimethoxy-1,3,5-triazin-2-yl)-4-methylmorpholinium chloride through coupling with N-methylmorpholine in tetrahydrofuran. Further, it plays an important role in the production of bis(4,6-dimethoxy-1,3,5-triazin-2-yl) ether, through reaction with 2-hydroxy-4,6-dimethoxy-1,3,5-triazine. In addition to this, it is involved in the preparation of 2-(4,6-dimethoxy-1,3,5-triazinyl)trialkylammonium salts through reaction with various tertiary amines.

Solubility
Soluble in water, chloroform and methanol.

Notes
Incompatible with strong oxidizing agents.
Melting Point 75°C to 79°C
Boiling Point 99°C to 100°C (1mmHg)
Quantity 5g
Beilstein 148988
Formula Weight 175.58
Percent Purity 98%
Chemical Name or Material 2-Chloro-4,6-dimethoxy-1,3,5-triazine

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning