Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4,6-dimethylpyridine-3-carbonitrile, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
14237-71-9
C8H7ClN2
166.61
MFCD00051676
RETJKTAVEQPNMH-UHFFFAOYSA-N
2-chloro-4,6-dimethylnicotinonitrile,2-chloro-3-cyano-4,6-dimethylpyridine,2-chloro-4,6-dimethyl-nicotinonitrile,2-chloro-4,6-dimethyl-3-pyridinecarbonitrile,3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl,acmc-1azio,chlorodimethylnicotinonitrile,ksc496m6f,2-chloro-4,6-dimethyinicotinonitrile,4,6-dimethyl-2-chloro-nicotinonitrile
223261
2-chloro-4,6-dimethylpyridine-3-carbonitrile
CC1=CC(=NC(=C1C#N)Cl)C
14237-71-9
C8H7ClN2
166.61
MFCD00051676
RETJKTAVEQPNMH-UHFFFAOYSA-N
2-chloro-4,6-dimethylnicotinonitrile,2-chloro-3-cyano-4,6-dimethylpyridine,2-chloro-4,6-dimethyl-nicotinonitrile,2-chloro-4,6-dimethyl-3-pyridinecarbonitrile,3-pyridinecarbonitrile, 2-chloro-4,6-dimethyl,acmc-1azio,chlorodimethylnicotinonitrile,ksc496m6f,2-chloro-4,6-dimethyinicotinonitrile,4,6-dimethyl-2-chloro-nicotinonitrile
223261
2-chloro-4,6-dimethylpyridine-3-carbonitrile
CC1=CC(=NC(=C1C#N)Cl)C
Quantity 5g
Melting Point 99.5°C to 101.0°C
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 166.61
Percent Purity 97%
Chemical Name or Material 2-chloro-4, 6-dimethylpyridine-3-carbonitrile

GHS H Statement:
Harmful if swallowed.
May cause respiratory irritation.
Harmful if inhaled.
Causes skin irritation.
Causes serious eye damage.
Harmful in contact with skin.

GHS P Statement:
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
Wear eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Immediately call a POISON CENTER or doctor/physician.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger