Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4,6-dimethylpyrimidine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
4472-44-0
C6H7ClN2
142.59
RZVPFDOTMFYQHR-UHFFFAOYSA-N
pyrimidine, 2-chloro-4,6-dimethyl,2-chloro-4,6-dimethyl-pyrimidine,pubchem6904,acmc-1adw7,5-23-05-00401 beilstein handbook reference,ksc495i5r,2-chlor-4,6-dimethylpyrimidine,4,6-dimethyl-2-chloropyrimidine,2-chloro-4,6-dimethyl-1,3-diazine,2-chloro-4,6-dimethylpyrimidine
20550
2-chloro-4,6-dimethylpyrimidine
CC1=CC(=NC(=N1)Cl)C
4472-44-0
C6H7ClN2
142.59
RZVPFDOTMFYQHR-UHFFFAOYSA-N
pyrimidine, 2-chloro-4,6-dimethyl,2-chloro-4,6-dimethyl-pyrimidine,pubchem6904,acmc-1adw7,5-23-05-00401 beilstein handbook reference,ksc495i5r,2-chlor-4,6-dimethylpyrimidine,4,6-dimethyl-2-chloropyrimidine,2-chloro-4,6-dimethyl-1,3-diazine,2-chloro-4,6-dimethylpyrimidine
20550
2-chloro-4,6-dimethylpyrimidine
CC1=CC(=NC(=N1)Cl)C
Boiling Point 210°C to 212°C
Melting Point 36°C to 38°C
Quantity 25g
Infrared Spectrum Authentic
Color White to Yellow
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 96% min. (GC)
Formula Weight 142.59
Physical Form Powder, Crystalline Powder or Low Melting Solid
Percent Purity 97%
Chemical Name or Material 2-Chloro-4, 6-dimethylpyrimidine
Show More Show Less

GHS H Statement
Harmful if swallowed.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.

GHS Signal Word: Warning