Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-aminopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
14432-12-3
C5H5ClN2
128.56
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
14432-12-3
C5H5ClN2
128.56
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
Melting Point 92°C to 94°C
Quantity 5g
Infrared Spectrum Authentic
Color Yellow
Packaging Glass bottle
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 128.56
Physical Form Crystalline Powder
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-aminopyridine
Show More Show Less

GHS H Statement
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breat

GHS Signal Word: Danger

EINECSNumber : 238-403-