Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-aminopyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
100g
Packaging:
Glass bottle
14432-12-3
C5H5ClN2
128.56
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
14432-12-3
C5H5ClN2
128.56
BLBDTBCGPHPIJK-UHFFFAOYSA-N
4-amino-2-chloropyridine,2-chloro-4-aminopyridine,2-chloro-pyridin-4-ylamine,4-pyridinamine, 2-chloro,2-chloro-4-pyridinamine,2-chlorpyridin-4-amin,4-amino-6-chloropyridine,2-chloro-4-pyridylamine,2-chloro-4-amino pyridine,4-amino-2-chloro pyridine
84432
2-chloropyridin-4-amine
C1=CN=C(C=C1N)Cl
Melting Point 92°C to 94°C
Color Yellow
Packaging Glass bottle
Quantity 100g
Infrared Spectrum Authentic
CAS Min % 96.0
CAS Max % 100.0
Assay Percent Range 97%
Formula Weight 128.56
Physical Form Crystalline Powder
Percent Purity ≥96.0%
Chemical Name or Material 2-Chloro-4-aminopyridine
Show More Show Less

GHS P Statement
Toxic in contact with skin.
Toxic if inhaled.
Causes serious eye irritation.
Toxic if swallowed.
Causes skin irritation.
May cause respiratory irritation.

GHS P Statement
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Wear protective gloves/protective clothing/eye protection/face protection.
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Gently wash with plenty of soap and water.
Immediately call a POISON CENTER or doctor/physician.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing. WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Danger