Alfa Aesar™ 2-Chloro-4-cyanobenzenesulfonyl chloride, 95%

Manufacturer: Alfa Aesar™ H50379MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH50379MD

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Chemical Identifiers

    CAS 254749-11-6
    Molecular Formula C7H3Cl2NO2S
    Formula Weight 236.08
    MDL Number MFCD00177089
    UN Number UN2923
    Chemical Name or Material 2-Chloro-4-cyanobenzenesulfonyl chloride

    Specifications

    Formula Weight 236.08
    Quantity 250mg
    Molecular Formula C7H3Cl2NO2S
    MDL Number MFCD00177089
    CAS 254749-11-6
    UN Number UN2923
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE SOLIDS, TOXIC, N.O.S.
    Melting Point 101° to 105°C
    Chemical Name or Material 2-Chloro-4-cyanobenzenesulfonyl chloride
    Assay 95%

    GHS H Statement
    H331-H314-H318-H302-H312
    Toxic if inhaled.
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit