Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-ethynylpyridine, 97%, Acros Organics™

Click to view available options
Quantity:
1g
5g
945717-09-9
C7H4ClN
137.57
DLEWWBKXDRBGRF-UHFFFAOYSA-N
2-chloro-4-ethynyl-pyridine
23436831
2-chloro-4-ethynylpyridine
C#CC1=CC(=NC=C1)Cl
945717-09-9
C7H4ClN
137.57
DLEWWBKXDRBGRF-UHFFFAOYSA-N
2-chloro-4-ethynyl-pyridine
23436831
2-chloro-4-ethynylpyridine
C#CC1=CC(=NC=C1)Cl
Packaging Glass Bottle
Infrared Spectrum Authentic
Quantity 1g
Boiling Point 373.0°C
Assay Percent Range 96% min. (HPLC)
Formula Weight 137.57
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-ethynylpyridine

GHS H Statement
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes.
Remove contact lenses,if present and easy to do.
Continue rinsing.

GHS Signal Word: Warning