Alfa Aesar™ 2-Chloro-4-fluoro-3-methylbenzoic acid, 97%

Manufacturer: Alfa Aesar™ H51879MD

Quantity 250mg

 View more versions of this product

Catalog No. AAH51879MD

  Check Availability
Add to Cart

    Chemical Identifiers

    Molecular Formula C8H6ClFO2
    Formula Weight 188.59
    MDL Number MFCD11846444
    Chemical Name or Material 2-Chloro-4-fluoro-3-methylbenzoic acid

    Specifications

    Formula Weight 188.59
    Molecular Formula C8H6ClFO2
    MDL Number MFCD11846444
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.
    Health Hazard 3 GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Melting Point 186° to 189°C
    Quantity 250mg
    Chemical Name or Material 2-Chloro-4-fluoro-3-methylbenzoic acid
    Assay 97%

    Description

    Applications
    It is used in the synthesis and Antibacterial Evaluation of 6-Amino-8-methylquinolones.

    Notes
    Store away from oxidizing agents. Keep the container tightly closed and place it in a cool, dry and well ventilated condition.

    GHS H Statement
    H315-H319-H335
    Causes skin irritation.
    Causes serious eye irritation.
    May cause respiratory irritation.

    GHS P Statement
    P261-P280-P305+P351+P338-P304+P340-P405-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit