Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Alfa Aesar™ 2'-Chloro-4'-fluoroacetanilide, 98%
Click to view available options
Quantity:
25g
100g
Specifications
Quantity | 25g |
Melting Point | 116°C to 118°C |
Beilstein | 2719131 |
Formula Weight | 187.6 |
Percent Purity | 98% |
Chemical Name or Material | 2'-Chloro-4'-fluoroacetanilide |
Safety and Handling
GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P261-P264b-P270-P271-P280-P301+P310-P302+P352-P304+P340-P311-P312-P330-P361-P363-P501c
H301+H311+H331
DOTInformation : Hazard Class: 6.1; Packaging Group: III
EINECSNumber : 000-000-0
TSCA : No
Recommended Storage : Ambient temperatures