Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4'-fluoroacetophenone, 99%

Click to view available options
Quantity:
25g
100g
500g
456-04-2
C8H6ClFO
172.583
MFCD00011652
UJZWJOQRSMOFMA-UHFFFAOYSA-N
2-chloro-4'-fluoroacetophenone,2-chloro-1-4-fluorophenyl ethanone,p-fluorophenacyl chloride,4-fluorophenacyl chloride,alpha-chloro-4-fluoroacetophenone,4-chloroacetyl-1-fluorobenzene,alpha-chloro-p-fluoroacetophenone,2-chloro-1-4-fluorophenyl ethan-1-one,ethanone, 2-chloro-1-4-fluorophenyl,2'-chloro-4-fluoroacetophenone
120248
2-chloro-1-(4-fluorophenyl)ethanone
C1=CC(=CC=C1C(=O)CCl)F
456-04-2
C8H6ClFO
172.583
MFCD00011652
UJZWJOQRSMOFMA-UHFFFAOYSA-N
2-chloro-4'-fluoroacetophenone,2-chloro-1-4-fluorophenyl ethanone,p-fluorophenacyl chloride,4-fluorophenacyl chloride,alpha-chloro-4-fluoroacetophenone,4-chloroacetyl-1-fluorobenzene,alpha-chloro-p-fluoroacetophenone,2-chloro-1-4-fluorophenyl ethan-1-one,ethanone, 2-chloro-1-4-fluorophenyl,2'-chloro-4-fluoroacetophenone
120248
2-chloro-1-(4-fluorophenyl)ethanone
C1=CC(=CC=C1C(=O)CCl)F
Applications
1-methyl-2-(4-methylthio)phenyl-4-(4-fluoro)phenylimidazole which was synthesized by condensing N-methyl-4-(methylthio)benzamidine with 2-chloro-4′-fluoroacetophenone.

Solubility
Insoluble in water.

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.
Melting Point 46°C to 50°C
Quantity 500g
Flash Point >110°C (230°F)
Boiling Point 247°C
UN Number UN3261
Beilstein 637860
Formula Weight 172.59
Percent Purity 99%
Chemical Name or Material 2-Chloro-4'-fluoroacetophenone

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.

RTECSNumber : AM6550000