Alfa Aesar™ 2-Chloro-4'-fluoroacetophenone, 99%

Manufacturer: Alfa Aesar™ B2218822

Quantity 100g

 View more versions of this product

Catalog No. AAB2218822

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 456-04-2
    Molecular Formula C8H6ClFO
    Formula Weight 172.59
    MDL Number MFCD00011652
    UN Number UN3261
    Beilstein 637860
    Synonym alpha-Chloro-4-fluoroacetophenone; 4-Fluorophenacyl chloride
    Chemical Name or Material 2-Chloro-4'-fluoroacetophenone

    Specifications

    Formula Weight 172.59
    MDL Number MFCD00011652
    UN Number UN3261
    Health Hazard 3 GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Boiling Point 247°C
    Melting Point 46° to 50°C
    Molecular Formula C8H6ClFO
    CAS 456-04-2
    Beilstein 637860
    Synonym alpha-Chloro-4-fluoroacetophenone; 4-Fluorophenacyl chloride
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    DOT Information Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE SOLID, ACIDIC, ORGANIC, N.O.S.
    RTECS Number AM6550000
    Flash Point >110°C (230°F)
    Quantity 100g
    Chemical Name or Material 2-Chloro-4'-fluoroacetophenone
    Assay 99%

    Description

    Applications
    1-methyl-2-(4-methylthio)phenyl-4-(4-fluoro)phenylimidazole which was synthesized by condensing N-methyl-4-(methylthio)benzamidine with 2-chloro-4 ft.-fluoroacetophenone.

    Solubility
    Insoluble in water.

    Notes
    Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Store away from strong oxidizing agents.

    GHS H Statement
    H314-H318
    Causes severe skin burns and eye damage.
    Causes serious eye damage.

    GHS P Statement
    P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit