Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-fluoroanisole, 97%

Click to view available options
Quantity:
5g
25g
2267-25-6
C7H6ClFO
160.572
MFCD00672965
RKCGJVGMRPKPNY-UHFFFAOYSA-N
2-chloro-4-fluoroanisole,benzene, 2-chloro-4-fluoro-1-methoxy,2-chloro-4-fluoro-1-methoxy-benzene,pubchem2658,acmc-1cnak,intermediates-zcf02609,ksc497i7l
2773580
2-chloro-4-fluoro-1-methoxybenzene
COC1=C(C=C(C=C1)F)Cl
2267-25-6
C7H6ClFO
160.572
MFCD00672965
RKCGJVGMRPKPNY-UHFFFAOYSA-N
2-chloro-4-fluoroanisole,benzene, 2-chloro-4-fluoro-1-methoxy,2-chloro-4-fluoro-1-methoxy-benzene,pubchem2658,acmc-1cnak,intermediates-zcf02609,ksc497i7l
2773580
2-chloro-4-fluoro-1-methoxybenzene
COC1=C(C=C(C=C1)F)Cl
Boiling Point 197°C to 200°C
Quantity 5g
Flash Point 76°C (170°F)
Formula Weight 160.58
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoroanisole

GHS H Statement
H315-H319-H335-H227
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Combustible liquid.

GHS P Statement
P210-P261-P280-P305+P351+P338-P405-P501a
Keep away from heat/sparks/open flames/hot surfaces. - No smoking.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning