Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-fluorobenzoyl chloride, 97%

Click to view available options
Quantity:
1g
25g
21900-54-9
C7H3Cl2FO
192.998
MFCD00084954
POIAZJJVWRVLBO-UHFFFAOYSA-N
2-chloro-4-fluorobenzoylchloride,2-chloro-4-fluoro-benzoyl chloride,2-chloro-4-fluorobenzene-1-carbonyl chloride,benzoyl chloride, 2-chloro-4-fluoro,benzoyl chloride,2-chloro-4-fluoro,acmc-20anzq,ksc547s3d,2-chlor-4-fluorbenzoylchlorid,2chloro-4-fluorobenzoyl chloride,4-fluoro-2-chlorobenzoyl chloride
2736546
2-chloro-4-fluorobenzoyl chloride
C1=CC(=C(C=C1F)Cl)C(=O)Cl
21900-54-9
C7H3Cl2FO
192.998
MFCD00084954
POIAZJJVWRVLBO-UHFFFAOYSA-N
2-chloro-4-fluorobenzoylchloride,2-chloro-4-fluoro-benzoyl chloride,2-chloro-4-fluorobenzene-1-carbonyl chloride,benzoyl chloride, 2-chloro-4-fluoro,benzoyl chloride,2-chloro-4-fluoro,acmc-20anzq,ksc547s3d,2-chlor-4-fluorbenzoylchlorid,2chloro-4-fluorobenzoyl chloride,4-fluoro-2-chlorobenzoyl chloride
2736546
2-chloro-4-fluorobenzoyl chloride
C1=CC(=C(C=C1F)Cl)C(=O)Cl
Quantity 1g
Boiling Point 82°C to 84°C (15mmHg)
UN Number UN3265
Beilstein 2254071
Formula Weight 193.01
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzoyl chloride

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

GHS P Statement
P260-P303+P361+P353-P305+P351+P338-P301+P330+P331-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF SWALLOWED: Rinse mouth. Do NOT induce vomiting.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 8; Packing Group: II; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.