Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-fluorobenzyl bromide, 97%

Click to view available options
Quantity:
5g
25g
45767-66-6
C7H5BrClF
223.469
MFCD00236025
GAUUDQVOPUKGJD-UHFFFAOYSA-N
2-chloro-4-fluorobenzyl bromide,1-bromomethyl-2-chloro-4-fluorobenzene,2-chloro-4-fluorobenzylbromide,alpha-bromo-2-chloro-4-fluorotoluene,benzene, 1-bromomethyl-2-chloro-4-fluoro,2-choro-4-fluorobenzylbromide,pubchem4893,1-bromomethyl-2-chloro-4-fluoro-benzene,ksc493m8r,2-chloro-4-flurobenzyl bromide
2725062
1-(bromomethyl)-2-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)Cl)CBr
45767-66-6
C7H5BrClF
223.469
MFCD00236025
GAUUDQVOPUKGJD-UHFFFAOYSA-N
2-chloro-4-fluorobenzyl bromide,1-bromomethyl-2-chloro-4-fluorobenzene,2-chloro-4-fluorobenzylbromide,alpha-bromo-2-chloro-4-fluorotoluene,benzene, 1-bromomethyl-2-chloro-4-fluoro,2-choro-4-fluorobenzylbromide,pubchem4893,1-bromomethyl-2-chloro-4-fluoro-benzene,ksc493m8r,2-chloro-4-flurobenzyl bromide
2725062
1-(bromomethyl)-2-chloro-4-fluorobenzene
C1=CC(=C(C=C1F)Cl)CBr
Melting Point 35°C to 39°C
Quantity 5g
UN Number UN3261
Beilstein 3539265
Formula Weight 223.48
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorobenzyl bromide

GHS H Statement
H314-H318
Causes severe skin burns and eye damage.
Causes serious eye damage.

P260-P264b-P270-P280-P303+P361+P353-P304+P340-P305+P351+P338-P310-P330-P331-P363-P501c

H302-H314

DOTInformation : Hazard Class: 8; Packaging Group: III

TSCA : No

Recommended Storage : Ambient temperatures