Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloro-4-fluorobenzyl Bromide 98.0+%, TCI America™
Click to view available options
Quantity:
5 g
25 g
Specifications
Melting Point | 39°C |
UN Number | 3261 |
Quantity | 25 g |
Formula Weight | 223.47 |
Percent Purity | ≥98.0% (GC) |
Physical Form | Crystalline Powder |
Chemical Name or Material | 2-Chloro-4-fluorobenzyl Bromide |
Safety and Handling
TSCA : No
Recommended Storage : Refrigerator