Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-fluorocinnamic acid, 97%

Click to view available options
Quantity:
1g
5g
25g
133220-86-7
C9H6ClFO2
200.593
MFCD00236027
RJCWBTRMWGOREZ-DUXPYHPUSA-N
2-chloro-4-fluorocinnamic acid,rarechem bk hw 0063,timtec-bb sbb003541,3-2-chloro-4-fluorophenyl-2-propenoic acid,2e-3-2-chloro-4-fluorophenyl prop-2-enoic acid,3-2-chloro-4-fluorophenyl acrylic acid,pubchem3429,e-3-2-chloro-4-fluorophenyl acrylic acid,3-chloro-5-fluorobenzoic,2-chloro-4-fluorocinnamicacid
6257297
(E)-3-(2-chloro-4-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)Cl)C=CC(=O)O
133220-86-7
C9H6ClFO2
200.593
MFCD00236027
RJCWBTRMWGOREZ-DUXPYHPUSA-N
2-chloro-4-fluorocinnamic acid,rarechem bk hw 0063,timtec-bb sbb003541,3-2-chloro-4-fluorophenyl-2-propenoic acid,2e-3-2-chloro-4-fluorophenyl prop-2-enoic acid,3-2-chloro-4-fluorophenyl acrylic acid,pubchem3429,e-3-2-chloro-4-fluorophenyl acrylic acid,3-chloro-5-fluorobenzoic,2-chloro-4-fluorocinnamicacid
6257297
(E)-3-(2-chloro-4-fluorophenyl)prop-2-enoic acid
C1=CC(=C(C=C1F)Cl)C=CC(=O)O
Melting Point 245°C to 249°C
Quantity 1g
Formula Weight 200.6
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluorocinnamic acid

GHS H Statement
H315-H319-H335
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
P261-P280-P305+P351+P338-P304+P340-P405-P501a
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Warning