Alfa Aesar™ 2-Chloro-4-fluorophenol, 98+%

Manufacturer: Alfa Aesar™ A1086422

Quantity 100g

 View more versions of this product

Catalog No. AAA1086422

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 1996-41-4
    Molecular Formula C6H4ClFO
    Formula Weight 146.55
    MDL Number MFCD00002168
    UN Number UN3265
    Beilstein 1861277
    Chemical Name or Material 2-Chloro-4-fluorophenol

    Specifications

    Formula Weight 146.55
    Health Hazard 3 GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.
    Molecular Formula C6H4ClFO
    CAS 1996-41-4
    Health Hazard 2 GHS H Statement
    H314-H318-H302-H312-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.
    Flash Point 75°C (167°F)
    UN Number UN3265
    Beilstein 1861277
    Health Hazard 1 Danger
    Odor Phenol-like
    Melting Point 20° to 23°C
    MDL Number MFCD00002168
    DOT Information Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, ORGANIC, N.O.S.
    Boiling Point 171° to 172°C
    Quantity 100g
    Chemical Name or Material 2-Chloro-4-fluorophenol
    Assay ≥98%

    GHS H Statement
    H314-H318-H302-H312-H227
    Causes severe skin burns and eye damage.
    Causes serious eye damage.
    Harmful if swallowed.
    Harmful in contact with skin.
    Combustible liquid.

    GHS P Statement
    P280-P273-P303+P361+P353-P305+P351+P338-P310
    Wear protective gloves/protective clothing/eye protection/face protection.
    Avoid release to the environment.
    IF ON SKIN (or hair): Remove/Take off immediately all contaminated clothing. Rinse skin with water/shower.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do.
    Continue rinsing.
    Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit