Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
2-Chloro-4-fluorophenylacetic acid, 97%, Thermo Scientific Chemicals
Click to view available options
Quantity:
5 g
25 g
Description
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
2-Chloro-4-fluorophenylacetic acid is used as a pharmaceutical intermediate.
Solubility
Slightly soluble in water.
Notes
Store in cool, dry conditions in a sealed container. Incompatible with oxidizing agents.
Specifications
Melting Point | 102°C to 105°C |
Quantity | 5 g |
Synonym | 2-chloro-4-fluorophenylacetic acid, 2-2-chloro-4-fluorophenyl acetic acid, 2-chloro-4-fluorophenyl acetic acid, 2-2-chloro-4-fluoro-phenyl acetic acid, rarechem al bo 0843, timtec-bb sbb003675, labotest-bb lt00441150, benzeneacetic acid,2-chloro-4-fluoro, 2-chloro-4-fluoro-phenyl-acetic acid, benzeneacetic acid, 2-chloro-4-fluoro |
Solubility Information | Slightly soluble in water. |
Formula Weight | 188.59 |
Percent Purity | 97% |
Chemical Name or Material | 2-Chloro-4-fluorophenylacetic acid |
Safety and Handling
Hazard Category | H315-H319-H335 |
Hazard Statement | GHS H Statement H315-H319-H335 Causes skin irritation. Causes serious eye irritation. May cause respiratory irritation. |
Precautionary Statement | P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P501c |
EINECSNumber | 000-000-0 |
TSCA | No |
Recommended Storage | Ambient temperatures |
RUO – Research Use Only
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction