Alfa Aesar™ 2-Chloro-4-fluorophenylacetonitrile, 97%

Manufacturer: Alfa Aesar™ B2313214

Quantity 25g

 View more versions of this product

Catalog No. AAB2313214

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 75279-56-0
    Molecular Formula C8H5ClFN
    Formula Weight 169.59
    MDL Number MFCD00236029
    UN Number UN3439
    Synonym 2-Chloro-4-fluorobenzyl cyanide
    Chemical Name or Material 2-Chloro-4-fluorophenylacetonitrile

    Specifications

    Formula Weight 169.59
    MDL Number MFCD00236029
    UN Number UN3439
    Melting Point 67° to 69°C
    Quantity 25g
    Molecular Formula C8H5ClFN
    CAS 75279-56-0
    Synonym 2-Chloro-4-fluorobenzyl cyanide
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H300-H311-H331-H315-H319
    Fatal if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.
    DOT Information Transport Hazard Class: 6.1; Packing Group: II; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Chemical Name or Material 2-Chloro-4-fluorophenylacetonitrile
    Assay 97%

    GHS H Statement
    H300-H311-H331-H315-H319
    Fatal if swallowed.
    Toxic in contact with skin.
    Toxic if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P280h-P305+P351+P338-P309-P310
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF exposed or if you feel unwell: Immediately call a POISON CENTER or doctor/physician.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit