Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-fluoropyridine, 97%, ACROS Organics™

Click to view available options
Quantity:
1g
5g
Packaging:
Glass bottle
34941-91-8
C5H3ClFN
131.54
MFCD04039345
FGSAQRJRWCZLOB-UHFFFAOYSA-N
pyridine, 2-chloro-4-fluoro,2-chloro-4-fluoro pyridine,2-chloro-4-fluoro-pyridine,pubchem6180,acmc-1aft7,2-chlro-4-fluoroxypyridine,ksc222e0n,pyrimidine, 4-chloro-6-fluoro,abbypharma ap-14-5731
2782792
2-chloro-4-fluoropyridine
C1=CN=C(C=C1F)Cl
34941-91-8
C5H3ClFN
131.54
MFCD04039345
FGSAQRJRWCZLOB-UHFFFAOYSA-N
pyridine, 2-chloro-4-fluoro,2-chloro-4-fluoro pyridine,2-chloro-4-fluoro-pyridine,pubchem6180,acmc-1aft7,2-chlro-4-fluoroxypyridine,ksc222e0n,pyrimidine, 4-chloro-6-fluoro,abbypharma ap-14-5731
2782792
2-chloro-4-fluoropyridine
C1=CN=C(C=C1F)Cl
Boiling Point 140°C
Flash Point 54°C
Specific Gravity 1.456
Density 1.4560g/mL
Quantity 1g
Color Colorless to Yellow
Packaging Glass bottle
Assay Percent Range 96% min. (GC)
Formula Weight 131.54
Physical Form Liquid
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-fluoropyridine
Show More Show Less

GHS H Statement
Flammable liquid and vapour.
Toxic if swallowed.
Toxic in contact with skin.
Toxic if inhaled.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.

GHS P Statement
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash

GHS Signal Word: Danger