Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-iodoaniline, 98%, ACROS Organics™

Click to view available options
Quantity:
1g
Packaging:
Glass bottle
42016-93-3
C6H5ClIN
253.467
MFCD00025298
MYDAOWXYGPEPJT-UHFFFAOYSA-N
2-chloro-4-iodo-phenylamine,benzenamine, 2-chloro-4-iodo,2-chloro-4-iodo-aniline,ccris 9438,zlchem 542,pubchem3811,2-chloro-4-iodoanilline,2-chloro-4-iodo aniline,acmc-1am5p,2-chloro-4-iodophenylamine
282930
2-chloro-4-iodoaniline
C1=CC(=C(C=C1I)Cl)N
42016-93-3
C6H5ClIN
253.467
MFCD00025298
MYDAOWXYGPEPJT-UHFFFAOYSA-N
2-chloro-4-iodo-phenylamine,benzenamine, 2-chloro-4-iodo,2-chloro-4-iodo-aniline,ccris 9438,zlchem 542,pubchem3811,2-chloro-4-iodoanilline,2-chloro-4-iodo aniline,acmc-1am5p,2-chloro-4-iodophenylamine
282930
2-chloro-4-iodoaniline
C1=CC(=C(C=C1I)Cl)N
Packaging Glass bottle
Quantity 1g
Melting Point 69°C to 73°C
Color Gray to Orange
Infrared Spectrum Authentic
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Linear Formula ClC6H3(I)NH2
Formula Weight 253.47
Physical Form Crystalline Powder
Percent Purity ≥97.5%
Chemical Name or Material 2-Chloro-4-iodoaniline, 98%
Show More Show Less

GHS H Statement:
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Harmful if inhaled.
Harmful in contact with skin.
May cause respiratory irritation.

GHS P Statement:
Wear protective gloves/protective clothing/eye protection/face protection.
IF ON SKIN: Wash with plenty of soap and water.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning

TSCA : TSCA