Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-iodopyridine, 98%, ACROS Organics™

Click to view available options
Quantity:
5g
25g
Packaging:
Glass bottle
153034-86-7
C5H3ClIN
239.44
MFCD01861983
KJKIPRQNFDUULB-UHFFFAOYSA-N
2-chloro-4-iodo-pyridine,4-iodo-2-chloropyridine,2-chloro-iodopyridine,pyridine, 2-chloro-4-iodo,zlchem 826,pubchem2579,acmc-209d8n,2-chloro-4-iodo pyridine,ksc174s0f,2-chloro-4-iodopyridine
1516511
2-chloro-4-iodopyridine
C1=CN=C(C=C1I)Cl
153034-86-7
C5H3ClIN
239.44
MFCD01861983
KJKIPRQNFDUULB-UHFFFAOYSA-N
2-chloro-4-iodo-pyridine,4-iodo-2-chloropyridine,2-chloro-iodopyridine,pyridine, 2-chloro-4-iodo,zlchem 826,pubchem2579,acmc-209d8n,2-chloro-4-iodo pyridine,ksc174s0f,2-chloro-4-iodopyridine
1516511
2-chloro-4-iodopyridine
C1=CN=C(C=C1I)Cl
Quantity 25g
Packaging Glass bottle
Infrared Spectrum Authentic
Color White to Yellow
Melting Point 41.0°C to 45.0°C
CAS Min % 97.5
CAS Max % 100.0
Assay Percent Range 98%
Formula Weight 239.44
Physical Form Crystals or Crystalline Flakes
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-iodopyridine
Show More Show Less

GHS H Statement:
Causes serious eye irritation.
May cause respiratory irritation.
Harmful if swallowed.
Causes skin irritation.
Harmful if inhaled.

GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF SWALLOWED: Call a POISON CENTER or doctor/physician if you feel unwell.
IF INHALED: Remove victim to fresh air and keep at rest in a position comfortable for breathing.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.

GHS Signal Word: Warning