Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-methoxyphenol, 97%, Thermo Scientific Chemicals

Catalog Number p-7023266
Click to view available options
Quantity:
5 g
25 g
18113-03-6
C7H7ClO2
158.581
MFCD00070773
GNVRRKLFFYSLGT-UHFFFAOYSA-N
87459
2-chloro-4-methoxyphenol
COC1=CC(=C(C=C1)O)Cl
18113-03-6
C7H7ClO2
158.581
MFCD00070773
GNVRRKLFFYSLGT-UHFFFAOYSA-N
87459
2-chloro-4-methoxyphenol
COC1=CC(=C(C=C1)O)Cl

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point 39°C to 44°C
Density 1.153
Flash Point 85°C (185°F)
Odor Phenol-like
Quantity 5 g
Beilstein 1936412
Synonym 3-chloro-4-hydroxyanisole, phenol, chloro-4-methoxy, phenol, 2-chloro-4-methoxy, pubchem3663, 2-chloro-4-methoxy-phenol, acmc-1c55n, phenol,2-chloro-4-methoxy, ksc445k0p, 2-chloro-4-methoxyphenol
Formula Weight 158.59
Percent Purity 97%
Chemical Name or Material 2-Chloro-4-methoxyphenol
Hazard Category H302+H312+H332-H315-H319-H335
Hazard Statement GHS H Statement
H302-H312-H315-H319-H335
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P261-P264b-P270-P271-P280-P301+P312-P302+P352-P304+P340-P305+P351+P338-P312-P330-P332+P313-P362-P501c
DOTInformation Hazard Class: 6.1; Packaging Group: III
EINECSNumber 242-007-3
TSCA No
Recommended Storage Ambient temperatures

RUO – Research Use Only

Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit