Alfa Aesar™ 2-Chloro-4-methoxyphenylacetonitrile, 95%

Manufacturer: Alfa Aesar™ H2602106

Quantity 5g

 View more versions of this product

Catalog No. AAH2602106

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 170737-93-6
    Molecular Formula C9H8ClNO
    Formula Weight 181.62
    MDL Number MFCD08445710
    UN Number UN3439
    Synonym 2-Chloro-4-methoxybenzyl cyanide
    Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile

    Specifications

    Formula Weight 181.62
    Synonym 2-Chloro-4-methoxybenzyl cyanide
    Health Hazard 1 Warning
    Health Hazard 2 GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: NITRILES, SOLID, TOXIC, N.O.S.
    Melting Point 41° to 43°C
    Molecular Formula C9H8ClNO
    MDL Number MFCD08445710
    CAS 170737-93-6
    UN Number UN3439
    Health Hazard 3 GHS P Statement
    P261-P280h-P305+P351+P338-P304+P340-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    Quantity 5g
    Chemical Name or Material 2-Chloro-4-methoxyphenylacetonitrile
    Assay 95%

    Description

    Applications
    2-Chloro-4-methoxyphenylacetonitrile is used as a raw material and an intermediate in organic synthesis.

    Solubility
    Insoluble in water.

    Notes
    Incompatible with strong oxidizing agents and acids.

    GHS H Statement
    H302-H312-H332-H315-H319
    Harmful if swallowed.
    Harmful in contact with skin.
    Harmful if inhaled.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P261-P280h-P305+P351+P338-P304+P340-P362-P501a
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/clothing.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    IF INHALED: Remove to fresh air and keep at rest in a position comfortable for breathing.
    Take off contaminated clothing and wash before reuse.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Warning

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit