Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Alfa Aesar™ 2-Chloro-4-methylaniline, 98%

Click to view available options
Quantity:
5g
25g
100g
615-65-6
C7H8ClN
141.598
MFCD00007666
XGYLSRFSXKAYCR-UHFFFAOYSA-N
2-chloro-p-toluidine,benzenamine, 2-chloro-4-methyl,4-amino-3-chlorotoluene,p-toluidine, 2-chloro,2-chlor-4-toluidin,4-methyl-2-chloroaniline,unii-k59isj2ko1,2-chlor-4-toluidin czech,ccris 2887,3-chloro-4-aminotoluene
12007
2-chloro-4-methylaniline
CC1=CC(=C(C=C1)N)Cl
615-65-6
C7H8ClN
141.598
MFCD00007666
XGYLSRFSXKAYCR-UHFFFAOYSA-N
2-chloro-p-toluidine,benzenamine, 2-chloro-4-methyl,4-amino-3-chlorotoluene,p-toluidine, 2-chloro,2-chlor-4-toluidin,4-methyl-2-chloroaniline,unii-k59isj2ko1,2-chlor-4-toluidin czech,ccris 2887,3-chloro-4-aminotoluene
12007
2-chloro-4-methylaniline
CC1=CC(=C(C=C1)N)Cl
Flash Point 99°C (210°F)
Boiling Point 223°C to 225°C
Quantity 5g
Melting Point 7°C
UN Number UN3429
Beilstein 774514
Formula Weight 141.6
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylaniline

GHS H Statement
H331-H373-H302-H312-H315-H319
Toxic if inhaled.
May cause damage to organs through prolonged or repeated exposure.
Harmful if swallowed.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

GHS P Statement
P260-P261-P280-P305+P351+P338-P405-P501a
Do not breathe dust/fume/gas/mist/vapors/spray.
Avoid breathing dust/fume/gas/mist/vapors/spray.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
Store locked up.
Dispose of contents/container in accordance with local/regional/national/international regulations.

Danger

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, LIQUID

RTECSNumber : XU5110000

TSCA : TSCA