Alfa Aesar™ 2-Chloro-4-methylaniline, 98%

Manufacturer: Alfa Aesar™ B22935-14

Quantity 25g

 View more versions of this product

Catalog No. AAB2293514

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 615-65-6
    Molecular Formula C7H8ClN
    Formula Weight 141.60g/mol
    MDL Number MFCD00007666
    UN Number UN3429
    Beilstein 774514
    Synonym 4-Amino-3-chlorotoluene; 2-Chloro-p-toluidine
    Chemical Name or Material 2-Chloro-4-methylaniline

    Specifications

    Melting Point 7°C
    Boiling Point 223° to 225°C
    Quantity 25g
    Flash Point 99°C (210°F)
    CAS 615-65-6
    Molecular Formula C7H8ClN
    Formula Weight 141.60g/mol
    MDL Number MFCD00007666
    UN Number UN3429
    Beilstein 774514
    Synonym 4-Amino-3-chlorotoluene; 2-Chloro-p-toluidine
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.
    Health Hazard 3 GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.
    DOT Information Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: CHLOROTOLUIDINES, LIQUID
    RTECS Number XU5110000
    TSCA TSCA
    Chemical Name or Material 2-Chloro-4-methylaniline
    Assay 98%

    GHS H Statement
    H331-H373-H302-H312-H315-H319
    Toxic if inhaled.
    May cause damage to organs through prolonged or repeated exposure.
    Harmful if swallowed.
    Harmful in contact with skin.
    Causes skin irritation.
    Causes serious eye irritation.

    GHS P Statement
    P260-P261-P280-P305+P351+P338-P405-P501a
    Do not breathe dust/fume/gas/mist/vapors/spray.
    Avoid breathing dust/fume/gas/mist/vapors/spray.
    Wear protective gloves/protective clothing/eye protection/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Store locked up.
    Dispose of contents/container in accordance with local/regional/national/international regulations.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit