Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

2-Chloro-4-methylpyrimidine, 98%, Acros Organics

Click to view available options
Quantity:
1g
10g
13036-57-2
C5H5ClN2
128.56
BHAKRVSCGILCEW-UHFFFAOYSA-N
2-chloro-4-methyl-pyrimidine,pyrimidine, 2-chloro-4-methyl,2-chloro-6-methylpyrimidine,pubchem6906,chloro-6 methylpyrimidine,4-methyl-2-chloropyrimidine,2-chloro-4-methyl pyrimidine,4-methyl-2-chloro-pyrimidine,ksc493s3l,4-methyl-2-pyrimidinylchloride
11629607
2-chloro-4-methylpyrimidine
CC1=NC(=NC=C1)Cl
13036-57-2
C5H5ClN2
128.56
BHAKRVSCGILCEW-UHFFFAOYSA-N
2-chloro-4-methyl-pyrimidine,pyrimidine, 2-chloro-4-methyl,2-chloro-6-methylpyrimidine,pubchem6906,chloro-6 methylpyrimidine,4-methyl-2-chloropyrimidine,2-chloro-4-methyl pyrimidine,4-methyl-2-chloro-pyrimidine,ksc493s3l,4-methyl-2-pyrimidinylchloride
11629607
2-chloro-4-methylpyrimidine
CC1=NC(=NC=C1)Cl
Boiling Point 93°C to 95°C (17.0mmHg)
Melting Point 51°C to 56°C
Quantity 10g
Assay Percent Range 97.5% min. (GC)
Formula Weight 128.56
Percent Purity 98%
Chemical Name or Material 2-Chloro-4-methylpyrimidine

GHS Signal Word: Warning