Alfa Aesar™ 2-Chloro-4-(methylsulfonyl)benzoic acid, 96%

Manufacturer: Alfa Aesar™ B2531903

Quantity 1g

 View more versions of this product

Catalog No. AAB2531903

  Check Availability
Add to Cart

    Chemical Identifiers

    CAS 53250-83-2
    Molecular Formula C8H7ClO4S
    Formula Weight 234.66
    MDL Number MFCD00216496
    Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid

    Specifications

    Formula Weight 234.66
    Molecular Formula C8H7ClO4S
    MDL Number MFCD00216496
    CAS 53250-83-2
    Health Hazard 1 Danger
    Health Hazard 2 GHS H Statement
    H318
    Causes serious eye damage.
    Health Hazard 3 GHS P Statement
    P280i-P305+P351+P338
    Wear eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
    Melting Point 191° to 196°C
    Quantity 1g
    Chemical Name or Material 2-Chloro-4-(methylsulfonyl)benzoic acid
    Assay 96%

    GHS H Statement
    H318
    Causes serious eye damage.

    GHS P Statement
    P280i-P305+P351+P338
    Wear eye/face protection.
    IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

    Danger

    SDS & Safety Documents

  • Description & Specifications
  • Safety and Handling

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit